TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade, Thermo Scientific Chemicals
TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade, Thermo Scientific Chemicals
TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade, Thermo Scientific Chemicals
TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals
TE Buffer, 1X Solution pH 8.0, Low EDTA, Molecular Biology Grade, Thermo Scientific Chemicals
Use directly to re-suspend and/or dilute purified DNA or RNA
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Nalgene bottle
Catalog number ALFJ75793.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Nalgene bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 1.5mm (0.06in) diameter.
Appearance (Color)Brown
Assay from Supplier's CofA≥99.999% (metals basis)
Use directly to re-suspend and/or dilute purified DNA or RNA.