Tris-HCl, 0.5M buffer soln., pH 8.0, low endotoxin
Tris-HCl, 0.5M buffer soln., pH 8.0, low endotoxin
Tris-HCl, 0.5M buffer soln., pH 8.0, low endotoxin
Tris-HCl, 0.5M buffer soln., pH 8.0, low endotoxin
Thermo Scientific Chemicals
Tris-HCl, 0.5M buffer soln., pH 8.0, low endotoxin
CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
Catalog number ALFJ67510.AK
Price (MYR)
903.00
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
903.00
Add to cart
Specifications
Chemical Name or MaterialTRIS-HCl
Name Notelow endotoxin
CAS1185-53-1