Gentamicin sulfate, 20 mg/ml in distilled water, sterile-filtered
Gentamicin sulfate, 20 mg/ml in distilled water, sterile-filtered
Gentamicin sulfate, 20 mg/ml in distilled water, sterile-filtered
Gentamicin sulfate, 20 mg/ml in distilled water, sterile-filtered
Thermo Scientific Chemicals
Gentamicin sulfate, 20 mg/ml in distilled water, sterile-filtered
CAS: 1405-41-0
Have Questions?
Catalog NumberQuantity
ALFJ67488.AD50 mL
Catalog number ALFJ67488.AD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS119-64-2
IUPAC Name1,2,3,4-tetrahydronaphthalene
Molecular FormulaC10H12
InChI KeyCXWXQJXEFPUFDZ-UHFFFAOYSA-N
SMILESC1CCC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.0 %
Appearance (Color)Clear colorless to light yellow
AppearanceMay darken on storage
Appearance (Form)Liquid