Gold nanoparticles, 15nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 520nm absorption, Thermo Scientific Chemicals
Gold nanoparticles, 15nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 520nm absorption, Thermo Scientific Chemicals
Gold nanoparticles, 15nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 520nm absorption, Thermo Scientific Chemicals
Gold nanoparticles, 15nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 520nm absorption, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gold nanoparticles, 15nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 520nm absorption, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFJ67307.AC25 mL
Catalog number ALFJ67307.AC
Price (MYR)
684.00
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (MYR)
684.00
Add to cart
Specifications
Chemical Name or MaterialGold nanoparticles
ColorPink to Red
Name Notesupplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 520nm absorption
CAS7440-57-5
Storage Note 12° to 8°C
View more
For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
RUO – Research Use Only