Boric acid, Electrophoresis Grade, 99.5% min, Thermo Scientific Chemicals
Boric acid, Electrophoresis Grade,  99.5% min, Thermo Scientific Chemicals
Boric acid, Electrophoresis Grade,  99.5% min, Thermo Scientific Chemicals
Boric acid, Electrophoresis Grade,  99.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boric acid, Electrophoresis Grade, 99.5% min, Thermo Scientific Chemicals
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Catalog number ALFJ67202.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS547-57-9
IUPAC Name4-[2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]benzene-1-sulfonic acid sodium
Molecular FormulaC12H10N2NaO5S
InChI KeyYOGUPPQYRWYKGT-UHFFFAOYSA-N
SMILES[Na].OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to brown