Boric acid, Electrophoresis Grade, 99.5% min, Thermo Scientific Chemicals
Boric acid, Electrophoresis Grade,  99.5% min, Thermo Scientific Chemicals
Boric acid, Electrophoresis Grade,  99.5% min, Thermo Scientific Chemicals
Boric acid, Electrophoresis Grade,  99.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boric acid, Electrophoresis Grade, 99.5% min, Thermo Scientific Chemicals
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Catalog number ALFJ67202.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS32864-38-3
IUPAC Name1-tert-butyl 3-ethyl propanedioate
Molecular FormulaC9H16O4
InChI KeyOCOBFMZGRJOSOU-UHFFFAOYSA-N
SMILESCCOC(=O)CC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4130-1.4190 @ 20?C
Assay (GC)≥94.0%