Amphotericin B, 250 μg/mL in distilled water, sterile-filtered
Amphotericin B, 250 μg/mL in distilled water, sterile-filtered
Amphotericin B, 250 μg/mL in distilled water, sterile-filtered
Amphotericin B, 250 μg/mL in distilled water, sterile-filtered
Thermo Scientific Chemicals
Amphotericin B, 250 μg/mL in distilled water, sterile-filtered
CAS: 1397-89-3 | C47H73NO17 | 924.09 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ67049.AD50 mL
Catalog number ALFJ67049.AD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS10101-63-0
IUPAC Nameλ²-lead(2+) diiodide
Molecular FormulaI2Pb
InChI KeyRQQRAHKHDFPBMC-UHFFFAOYSA-L
SMILES[I-].[I-].[Pb++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.5% min