Neomycin sulfate, 50 mg/ml in distilled water, sterile-filtered
Neomycin sulfate, 50 mg/ml in distilled water, sterile-filtered
Neomycin sulfate, 50 mg/ml in distilled water, sterile-filtered
Neomycin sulfate, 50 mg/ml in distilled water, sterile-filtered
Thermo Scientific Chemicals
Neomycin sulfate, 50 mg/ml in distilled water, sterile-filtered
CAS: 1405-10-3 | C23H48N6O17S | 712.72 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ67011.AD50 mL
Catalog number ALFJ67011.AD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS10025-65-7
IUPAC Nameplatinum(2+) dichloride
Molecular FormulaCl2Pt
InChI KeyCLSUSRZJUQMOHH-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pt++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Grey-green to brown
Assay>=71.8 % (Pt)