Cefuroxime sodium salt, 95%, Thermo Scientific Chemicals
Cefuroxime sodium salt, 95%, Thermo Scientific Chemicals
Cefuroxime sodium salt, 95%, Thermo Scientific Chemicals
Cefuroxime sodium salt, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cefuroxime sodium salt, 95%, Thermo Scientific Chemicals
CAS: 56238-63-2 | C16H15N4NaO8S | 446.366 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66999.031 g
Catalog number ALFJ66999.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCefuroxime sodium salt
TypeCefuroxime Sodium Salt
Physical FormPowder
CAS56238-63-2
Health Hazard 1H317-H334