Hyaluronic acid sodium salt, Streptococcus equi, 91%, Thermo Scientific Chemicals
Hyaluronic acid sodium salt, Streptococcus equi, 91%, Thermo Scientific Chemicals
Hyaluronic acid sodium salt, Streptococcus equi, 91%, Thermo Scientific Chemicals
Hyaluronic acid sodium salt, Streptococcus equi, 91%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hyaluronic acid sodium salt, Streptococcus equi, 91%, Thermo Scientific Chemicals
Hyaluronic acid sodium salt, CAS # 9067-32-7, is an acidic mucopolysaccharide produced by the bacterium Streptococcus zooepidemicus that is abundant in the mammalian extracellular matrix.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
Catalog number ALFJ66993.ME
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS7790-28-5
IUPAC Namesodium periodate
Molecular FormulaINaO4
InChI KeyJQWHASGSAFIOCM-UHFFFAOYSA-M
SMILES[Na+].[O-][I](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (special)≥97.5%
Bacterial hyaluronic acid (HA) is a natural non-sulfated high molecular weight glycosaminoglycan that may be used to study formulations of non-animal gels potentially useful for procedures such as soft tissue augmentation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Hyaluronic acid sodium salt is a component of the extracellular matrix that is potentially useful for soft tissue augmentation procedures
Application
• Hyaluronic acid sodium salt may be used in formulations of non-animal gels used in soft tissue augmentation studies
• It can be used in laboratory development of artificial synovial fluid
• It can also be used to study its role in oxidative stress and tumor progression
RUO – Research Use Only
General References:
  1. Barry Halliwell. Superoxide-dependent formation of hydroxyl radicals in the presence of iron salts: Its role in degradation of hyaluronic acid by a superoxide-generating system. FEBS Lett. 1978, 96 (2),238-242.