Cefoxitin, 98%
Cefoxitin, 98%
Cefoxitin, 98%
Thermo Scientific Chemicals
Cefoxitin, 98%
CAS: 35607-66-0 | C16H17N3O7S2 | 427.446 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66891.031 g
Catalog number ALFJ66891.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS18583-60-3
IUPAC Namepotassium tris(1H-pyrazol-1-yl)boranuide
Molecular FormulaC9H10BKN6
InChI KeyIBNGZWIUPDMZMG-UHFFFAOYSA-N
SMILES[K+].C1=CN(N=C1)[BH-](N1C=CC=N1)N1C=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO4>=91 %
Infrared spectrumConforms
Melting point182°C to 188°C
Appearance (Color)White
Cefoxitin is a semisynthetic, broad-spectrum second-generation cephalosporin with antibacterial activity. The activity of cefoxitin results in the weakening of the bacterial cell wall and causes cell lysis. Cefoxitin acts by interfering with cell wall synthesis. Its activity spectrum includes a broad range of gram-negative and gram-positive bacteria including anaerobes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cefoxitin is a semisynthetic, broad-spectrum second-generation cephalosporin with antibacterial activity. The activity of cefoxitin results in the weakening of the bacterial cell wall and causes cell lysis. Cefoxitin acts by interfe
RUO – Research Use Only
H. Knothe.; P. Shah.; V. Krcmery.; M. Antal.; S. Mitsuhashi. Transferable resistance to cefotaxime, cefoxitin, cefamandole and cefuroxime in clinical isolates of Klebsiella pneumoniae and Serratia marcescens.Infection. 1983, 11 (6),315-317 .