Cefoxitin sodium, 92.7-97%, Thermo Scientific Chemicals
Cefoxitin sodium, 92.7-97%, Thermo Scientific Chemicals
Cefoxitin sodium, 92.7-97%, Thermo Scientific Chemicals
Cefoxitin sodium, 92.7-97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cefoxitin sodium, 92.7-97%, Thermo Scientific Chemicals
A broad spectrum antibiotic
Have Questions?
Catalog NumberQuantity
ALFJ66800.031 g
Catalog number ALFJ66800.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCefoxitin sodium
TypeCefoxitin Sodium
Physical FormPowder
CAS33564-30-6
Health Hazard 1H317
View more
A broad spectrum antibiotic