Propidium iodide
Propidium iodide
Propidium iodide
Thermo Scientific Chemicals
Propidium iodide
CAS: 25535-16-4 | C27H34I2N4 | 668.406 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66764.MC100 mg
Catalog number ALFJ66764.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS338454-80-1
IUPAC Name4-iodo-2-methoxyaniline
Molecular FormulaC7H8INO
InChI KeyAEPCMLLYVXZOLQ-UHFFFAOYSA-N
SMILESCOC1=C(N)C=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Appearance (Form)Solid
Infrared spectrumConforms
GC>=96.0 %
NMR (1H-NMR)Consistent with structure
Propidium iodide is a fluorescent stain for nucleic acids. Cell membrane integrity excludes propidium iodide from staining viable and apoptotic cells. Propidium iodide may be used in flow cytometry to evaluate cell viability when used with other dyes that stain viable cells or cells that are early in the apoptosis process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Propidium iodide is a fluorescent stain for nucleic acids. Cell membrane integrity excludes propidium iodide from staining viable and apoptotic cells. Propidium iodide may be used in flow cytometry to evaluate cell viability when us
RUO – Research Use Only
General References:
  1. Bi-Ying Weia.; Yu-Min Juanga.; Chien-Chen Lai. A novel approach for quantitative peptides analysis by selected electron transfer reaction monitoring. Journal of Chromatography A. 2010, 1217, (44), 6927-6931.
  2. Hsuan-shen Chen.; Tomas Rejtar.; Viktor Andreev.; Eugene Moskoves.; Barry L. Karge. High-Speed, High-Resolution Monolithic Capillary LC-MALDI MS Using an Off-Line Continuous Deposition Interface for Proteomic Analysis. Anal. Chem. 2005, 77, (8), 2323-2331.