Sucralose, 98%
Sucralose, 98%
Sucralose, 98%
Thermo Scientific Chemicals
Sucralose, 98%
CAS: 56038-13-2 | C12H19Cl3O8 | 397.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
50 g
250 g
Catalog number ALFJ66736.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSucralose
CAS56038-13-2
Melting Point130°C (decomposition)
ColorWhite
Recommended StorageAmbient temperatures
View more
Sucralose is a non-caloric sugar used as a sweetener that activates T1R2/T1R3 sweet taste receptors and modulates glucagon-like peptide-1, glucose-dependent insulinotrophic peptide, and incretin secretion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sucralose is a non-caloric sugar used as a sweetener that activates T1R2/T1R3 sweet taste receptors and modulates glucagon-like peptide-1, glucose-dependent insulinotrophic peptide, and incretin secretion.
Solubility
Soluble in Water.
Notes
Research use only: Not intended for food, cosmetic, or diagnostic and therapeutic use in animals nor humans.
RUO – Research Use Only
General References:
  1. Goldsmith, L.A. Acute and subchronic toxicity of sucralose. Food and Chemical Toxicology. 2000, 38, (2), 53-69.
  2. Imma Ferrer.; Michael Thurman, E. Analysis of sucralose and other sweeteners in water and beverage samples by liquid chromatography/time-of-flight mass spectrometry. Journal of Chromatography A. 2010, 1217, (25), 4127-4134.