Picloram, 95%, Thermo Scientific Chemicals
Picloram, 95%, Thermo Scientific Chemicals
Picloram, 95%, Thermo Scientific Chemicals
Picloram, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Picloram, 95%, Thermo Scientific Chemicals
Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA)
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFJ66702.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS86087-24-3
IUPAC Nameoxolan-3-ol
Molecular FormulaC4H8O2
InChI KeyXDPCNPCKDGQBAN-UHFFFAOYNA-N
SMILESOC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=97.5 %
Specific optical rotation-15.0° to -21.0° (20°C, 589 nm) (c=2.4, methanol)
Appearance (Form)Liquid
Infrared spectrumConforms
Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA)