Ticarcillin disodium and clavulanate potassium (15:1)
Ticarcillin disodium and clavulanate potassium (15:1)
Ticarcillin disodium and clavulanate potassium (15:1)
Ticarcillin disodium and clavulanate potassium (15:1)
Thermo Scientific Chemicals
Ticarcillin disodium and clavulanate potassium (15:1)
CAS: 86482-18-0 | C23H25N3O11S2 | 583.583 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ALFJ66659.0910 gPlastic bottle
Catalog number ALFJ66659.09
Price (MYR)
1,176.00
Add to cart
Quantity:
10 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,176.00
Add to cart
Specifications
Chemical Name or MaterialTicarcillin disodium and clavulanate potassium (15:1)
CAS86482-18-0
Health Hazard 1H317-H334
Health Hazard 3P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
Recommended StorageKeep cold
View more
Combination provides greater efficacy against beta--lactam resistant strains