Dimethyl sulfoxide, Bioreagent, Thermo Scientific Chemicals
Dimethyl sulfoxide, Bioreagent, Thermo Scientific Chemicals
Dimethyl sulfoxide, Bioreagent, Thermo Scientific Chemicals
Dimethyl sulfoxide, Bioreagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl sulfoxide, Bioreagent, Thermo Scientific Chemicals
CAS: 67-68-5 | C2H6OS | 78.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
50 mL
250 mL
500 mL
Catalog number ALFJ66650.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS10111-08-7
IUPAC Name1H-imidazole-2-carbaldehyde
Molecular FormulaC4H4N2O
InChI KeyXYHKNCXZYYTLRG-UHFFFAOYSA-N
SMILESO=CC1=NC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to dark cream to cream to yellow to orange to brown
Assay (HPLC)≥96.0%
FormCrystals or powder or crystalline powder or lumps
DMSO is a polar aprotic solvent used in chemical reactions, in polymerase chain reactions and as a cryoprotectant vitrification agent for the preservation of cells, tissues and organs. DMSO is used in cell freezing media to protect cells from ice crystal induced mechanical injury. It is used for frozen storage of primary, sub-cultured, and recombinant heteroploid and hybridoma cell lines, embryonic stem cells and hematopoietic stem cells. DMSO is frequently used in the combinations with BSA or fetal bovine serum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DMSO is a polar aprotic solvent used in chemical reactions, in polymerase chain reactions and as a cryoprotectant vitrification agent for the preservation of cells, tissues and organs. DMSO is used in cell freezing media to protect cells from ice crystal induced mechanical injury. It is used for frozen storage of primary, sub-cultured, and recombinant heteroploid and hybridoma cell lines, embryonic stem cells and hematopoietic stem cells. DMSO is frequently used in the combinations with BSA or fetal bovine serum.
Solubility
Soluble in water, methanol, acetone, ether, benzene, chloroform.
Notes
Submicron filtered. Store at Room Temperature (15-30°C), desiccate. Hygroscopic.
RUO – Research Use Only