Riboflavin, 97%, Electrophoresis Grade
Riboflavin, 97%, Electrophoresis Grade
Riboflavin, 97%, Electrophoresis Grade
Riboflavin, 97%, Electrophoresis Grade
Thermo Scientific Chemicals
Riboflavin, 97%, Electrophoresis Grade
CAS: 83-88-5 | C17H20N4O6 | 376.37 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66632.1425 g
Catalog number ALFJ66632.14
Price (MYR)
504.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
504.00
Add to cart
Specifications
Chemical Name or MaterialRiboflavin
CAS83-88-5
Melting Point290°C
Storage Note 12° to 8°C
ColorOrange-Yellow