Triton∣r X-100, Electrophoresis Reagent
Triton∣r X-100, Electrophoresis Reagent
Triton∣r X-100, Electrophoresis Reagent
Triton∣r X-100, Electrophoresis Reagent
Thermo Scientific Chemicals
Triton∣r X-100, Electrophoresis Reagent
CAS: 9002-93-1 | C16H26O2, C16H26O2 | 250.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2500 mL
Catalog number ALFJ66624.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS65-85-0
IUPAC Namebenzoic acid
Molecular FormulaC7H6O2
InChI KeyWPYMKLBDIGXBTP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point121°C to 123°C
Sulfated ash=<0.01 %
Iron (Fe)=<4 ppm
Appearance (Form)Crystalline powder
Heavy metals (as Pb)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Ambient temperatures