Propidium iodide, 1mg/ml aqueous soln.
Propidium iodide, 1mg/ml aqueous soln.
Propidium iodide, 1mg/ml aqueous soln.
Propidium iodide, 1mg/ml aqueous soln.
Thermo Scientific Chemicals
Propidium iodide, 1mg/ml aqueous soln.
CAS: 25535-16-4 | C27H34I2N4 | 668.406 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66584.AB10 mL
Catalog number ALFJ66584.AB
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPropidium iodide
Name Note1mg/mL aqueous soln.
CAS25535-16-4
Storage Note 12° to 8°C
ColorOrange-Red
View more
Membrane impermeant, phenanthridinium intercalator dye excited by mercury or xenon-arc lamps or argon-ion laser. Selectively stains dead cells and a counter stain for multicolor fluorescent images.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Membrane impermeant, phenanthridinium intercalator dye excited by mercury or xenon-arc lamps or argon-ion laser. Selectively stains dead cells and a counter stain for multicolor fluorescent images.
Solubility
Diffic
RUO – Research Use Only
General References:
  1. Jeffrey D. Macklis.; Roger D. Madison. Progressive incorporation of propidium iodide in cultured mouse neurons correlates with declining electrophysiological status: a fluorescence scale of membrane integrity. Journal of Neuroscience Methods. 1990, 31, (1), 43-46.
  2. Chang-Jing G. Yeh.; Bae-Li Hsi.; W. Page Faulk. Propidium iodide as a nuclear marker in immunofluorescence. II. Use with cellular identification and viability studies. Journal of Immunological Methods. 1981, 43, (3), 269-275.