Sulfamethoxazole, 98%
Sulfamethoxazole, 98%
Sulfamethoxazole, 98%
Sulfamethoxazole, 98%
Thermo Scientific Chemicals
Sulfamethoxazole, 98%
Sulfamethoxazole, CAS # 723-46-6, is a sulfonamide antibiotic that inhibits folic acid synthesis in bacteria and prevents cell division and growth. | CAS: 723-46-6 | C10H11N3O3S | 253.276 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ALFJ66565.22100 gPlastic bottle
Catalog number ALFJ66565.22
Price (MYR)
1,171.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,171.00
Add to cart
Specifications
Chemical Name or MaterialSulfamethoxazole
CAS723-46-6
Health Hazard 1H302-H315-H319-H335
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
Melting Point168°C to 172°C
View more
A broad spectrum bacteriostatic sulfonamide antibiotic
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • Sulfamethoxazole is an antibiotic composed of an isoxazole with a 4-aminobenzenesulfonamido and methyl-group substitution at positions 5 and 3, respectively
  • Sulfamethoxazole carries out most of its action through the inhibition of dihydropteroate synthase, the enzyme responsible for the synthesis of the precursor of folic acid; inhibition of folic acid synthesis prevents DNA replication and cell division in susceptible bacteria
Application
  • Sulfamethoxazole can elicit an allergic response and may be useful for immune cell studies in laboratory settings
  • Sulfamethoxazole is suitable for laboratory research on infection, environmental pollution, and the physiology of susceptible bacteria
RUO – Research Use Only
General References:
  1. Xiong JQ, Govindwar S, Kurade MB, Paeng KJ, Roh HS, Khan MA, Jeon BH. Toxicity of sulfamethazine and sulfamethoxazole and their removal by a green microalga, Scenedesmus obliquus. Chemosphere. 2019 Mar;218:551-558. doi: 10.1016/j.chemosphere.2018.11.146. Epub 2018 Nov 24. PMID: 30500716.
  2. Ma R, Wang Y, Zou X, Hu K, Sun B, Fang W, Fu G, Yang X. Pharmacokinetics of sulfamethoxazole and trimethoprim in Pacific white shrimp, Litopenaeus vannamei, after oral administration of single-dose and multiple-dose. Environ Toxicol Pharmacol. 2017 Jun;52:90-98. doi: 10.1016/j.etap.2017.03.019. Epub 2017 Mar 28. PMID: 28410468.