Agarose, Electrophoresis Grade
Agarose, Electrophoresis Grade
Agarose, Electrophoresis Grade
Agarose, Electrophoresis Grade
Thermo Scientific Chemicals
Agarose, Electrophoresis Grade
CAS: 9012-36-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALFJ66501.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS608-66-2
IUPAC Namehexane-1,2,3,4,5,6-hexol
Molecular FormulaC6H14O6
InChI KeyFBPFZTCFMRRESA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Melting point187°C to 191°C
HPLC>=99.0 %
View more
Standard agarose suitable for most general applications. Good gel strength and clarity. Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Standard agarose suitable for most general applications. Good gel strength and clarity. Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it.
Solubility
Soluble in water(10g/L).
Notes
Gel strength (1.5% w/v): >1120g/cm^2; Gelling temperature: 36-39°C; EEO: <0.12
RUO – Research Use Only
General References:
  1. James M. Bailey.; Norman Davidson. Methylmercury as a reversible denaturing agent for agarose gel electrophoresis. Analytical Biochemistry. 1976, 70, (1), 75-85.
  2. Carl-Bertil Laurell. Quantitative estimation of proteins by electrophoresis in agarose gel containing antibodies. Analytical Biochemistry. 1966, 15, (1), 45-52.