Oxacillin sodium salt monohydrate, 815^mg/mg
Oxacillin sodium salt monohydrate, 815^mg/mg
Oxacillin sodium salt monohydrate, 815^mg/mg
Oxacillin sodium salt monohydrate, 815^mg/mg
Thermo Scientific Chemicals
Oxacillin sodium salt monohydrate, 815^mg/mg
CAS: 7240-38-2 | C19H23N3NaO6S | 444.458 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66380.031 g
Catalog number ALFJ66380.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.03% max.
Used primarily against Gram-positive organisms namely resistant Staphylococcus species
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Used primarily against gram positive organisms namely resistant Staphylococcus species
RUO – Research Use Only