Nisin, 900 I.U./mg, Thermo Scientific Chemicals
Nisin, 900 I.U./mg, Thermo Scientific Chemicals
Nisin, 900 I.U./mg, Thermo Scientific Chemicals
Nisin, 900 I.U./mg, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nisin, 900 I.U./mg, Thermo Scientific Chemicals
Nisin, CAS # 1414-45-5, is a polycyclic peptide derived from and produced by Lactococcus lactis that shows antibacterial activity against gram-positive bacteria.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALFJ66370.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNisin
TypeNisin
Name Note900 I.U./mg
Physical FormPowder
CAS1414-45-5
View more
Nisin, 900 I.U./mg is commonly used as a food preservative. It is also used as a selective agent in cell culture for the isolation of gram-negative bacteria, yeast, and molds. Nisin has been used in bioassays to study posttranslational modification. It binds and interferes with the permeability of the cytoplasmic membrane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • Nisin is a broad-spectrum polycyclic peptide derived from and produced by Lactococcus lactis that shows antibacterial activity against gram-positive bacteria Nisin binds lipid II in the cell membrane. After binding it can insert into the membrane, creating a pore Nisin can efficiently inhibit the growth of gram-positive bacteria, and resistance is rarely observed
Application
  • Nisin is commonly used as a selective compound in cell culture to isolate gram-negative bacteria, yeast, and molds (a type of fungus)
  • It has been used in bioassays to study posttranslational modification
  • This compound has the ability to bind and interfere with the permeability of the cytoplasmic membrane
  • It is used to study the assembly and stability of nisin-lipid II pore complexes
  • This compound is used in in vitro experiments to form targeted pores in cell membranes that are mediated by lipid II
RUO – Research Use Only
General References:
  1. Olli Koponen; Marja Tolonen; Mingqiang Qiao; Gudrun Wahlström; Jari Helin; Per E J Saris. NisB is required for the dehydration and NisC for the lanthionine formation in the post-translational modification of nisin. Microbiology.2002, 148, (Pt 11), 3561-3568.
  2. Brötz H.; and Sahl H. New insights into the mechanism of action of lantibiotics - Diverse biological effects by binding to the same molecular target. J Antimicrob Chemother.2000, 46, (1), 1-6.
  3. Hasper HE, de Kruijff B, Breukink E. Assembly and stability of nisin-lipid II pores. Biochemistry. 2004, 43, (36), 11567-11575.
  4. Wiedemann I, Benz R, Sahl HG. Lipid II-mediated pore formation by the peptide antibiotic nisin: a black lipid membrane study. J Bacteriol. 2004, 186, (10), 3259-3261.
  5. Zhou L, van Heel AJ, Montalban-Lopez M, Kuipers OP. Potentiating the Activity of Nisin against Escherichia coli. Front Cell Dev Biol. 2016, 4, 7.
  6. Olli Koponen; Marja Tolonen; Mingqiang Qiao; Gudrun Wahlström; Jari Helin; Per E J Saris. NisB is required for the dehydration and NisC for the lanthionine formation in the post-translational modification of nisin. Microbiology. 2002, 148, (Pt 11), 3561-3568.
  7. Brötz H.; and Sahl H. New insights into the mechanism of action of lantibiotics - Diverse biological effects by binding to the same molecular target. J Antimicrob Chemother. 2000, 46 (1), 1-6.