Polysorbate 20, Electrophoresis Reagent, Thermo Scientific Chemicals
Polysorbate 20, Electrophoresis Reagent, Thermo Scientific Chemicals
Polysorbate 20, Electrophoresis Reagent, Thermo Scientific Chemicals
Polysorbate 20, Electrophoresis Reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 20, Electrophoresis Reagent, Thermo Scientific Chemicals
CAS: 9005-64-5 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | 522.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number ALFJ66278.AE
Price (MYR)
249.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
249.00
Add to cart
Specifications
Chemical Name or MaterialPolysorbate 20
ColorColorless to Yellow
Name NoteElectrophoresis Reagent
CAS9005-64-5
Physical FormLiquid