Agarose, Genetic Technology Grade
Agarose, Genetic Technology Grade
Agarose, Genetic Technology Grade
Agarose, Genetic Technology Grade
Thermo Scientific Chemicals
Agarose, Genetic Technology Grade
CAS: 9012-36-6
Have Questions?
Catalog NumberQuantity
ALFJ66208.22100 g
Catalog number ALFJ66208.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAgarose
Physical FormPowder
CAS9012-36-6
Recommended StorageAmbient temperatures
ColorWhite
View more
Engineered to permit recovery of active DNA after separation, with minimal inhibition of restriction and ligation enzymes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Engineered to permit recovery of active DNA after separation, with minimal inhibition of restriction and ligation enzymes
RUO – Research Use Only