Glufosinate ammonium, 95%, Thermo Scientific Chemicals
Glufosinate ammonium, 95%, Thermo Scientific Chemicals
Glufosinate ammonium, 95%, Thermo Scientific Chemicals
Glufosinate ammonium, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glufosinate ammonium, 95%, Thermo Scientific Chemicals
Glufosinate ammonium, 95%, CAS # 77182-82-2, also known as glufosinate, is a non-selective phosphorous herbicide.
Have Questions?
Catalog NumberQuantity
ALFJ66186.031 g
Catalog number ALFJ66186.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS873-75-6
IUPAC Name(4-bromophenyl)methanol
Molecular FormulaC7H7BrO
InChI KeyVEDDBHYQWFOITD-UHFFFAOYSA-N
SMILESOCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point75°C to 78°C
Appearance (Color)White to light beige
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder or crystals
Used as broad spectrum post-emergence agrochemical for grapes, orchards, plantations, ornamentals, and non-cropland; Also used as crop desiccant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Glufosinate ammonium is a compound which exhibits broad-spectrum herbicidal properties through inhibition of glutamine synthetase, which disrupts cell membranes
Application
• Glufosinate ammonium acts as a non-selective herbicide
RUO – Research Use Only
General References:
  1. Griet Den Herder et. al. Seven in absentia proteins affect plant growth and nodulation in Medicago truncatula.. Plant Physiology., 2008,148(1), 369-382.
  2. Mahbod Sahebi; Mohamed M. Hanafi; A. Siti Nor Akmar; Mohd Y. Rafii; Parisa Azizi; A.S. Idris. Serine-rich protein is a novel positive regulator for silicon accumulation in mangrove. Gene, 2015,556(2), 170-181.