Acrylamide, 99.5%, Molecular Biology Grade
Acrylamide, 99.5%, Molecular Biology Grade
Acrylamide, 99.5%, Molecular Biology Grade
Acrylamide, 99.5%, Molecular Biology Grade
Thermo Scientific Chemicals
Acrylamide, 99.5%, Molecular Biology Grade
CAS: 79-06-1 | C3H5NO | 71.08 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66184.36500 g
Catalog number ALFJ66184.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS19583-77-8
IUPAC Namedisodium hexachloroplatinum hexahydrate
Molecular FormulaCl6H12Na2O6Pt
InChI KeyXGFSWVNCHVSCSP-UHFFFAOYSA-H
SMILESO.O.O.O.O.O.[Na+].[Na+].Cl[Pt](Cl)(Cl)(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Assay from Supplier's CofA31.30-37.00% w/w Platinum
FormCrystals
Acrylamide solution is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Also used for preparing slab gel gradients for PAGE.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acrylamide solution is a commonly used matrix for electrophoretic separation of nucleic acids and proteins. Also used for preparing slab gel gradients for PAGE.
Solubility
Partly soluble in water.
Notes
Air and Light Sensitive, store in dark and away form light. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Charles Beauchamp; Irwin Fridovich. Superoxide dismutase: Improved assays and an assay applicable to acrylamide gels. Analytical Biochemistry., 1971,44(1), 276-287.
  2. Robert M. Zacharius; Tatiana E. Zell; John H. Morrison; John J. Woodlock. Glycoprotein staining following electrophoresis on acrylamide gels. Analytical Biochemistry, 1969,30(1), 148-152.