Ascorbate oxidase, from Cucurbita sp.
Ascorbate oxidase, from Cucurbita sp.
Thermo Scientific Chemicals
Ascorbate oxidase, from Cucurbita sp.
Have Questions?
Catalog NumberQuantity
ALFJ65782.4I1 kU
Catalog number ALFJ65782.4I
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kU
Request bulk or custom format
Chemical Identifiers
CAS19438-10-9
IUPAC Namemethyl 3-hydroxybenzoate
Molecular FormulaC8H8O3
InChI KeyYKUCHDXIBAQWSF-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale gray
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)67.0-75.0°C
Ascorbate oxidase, from Cucurbita sp., may be used to study oxidative stress and heat stress response and tolerance. Also used in ascorbic acid assays to study the heat stress response of Arabidopsis and useful for enzymatic determination of ascorbic acid and for eliminating the interference of ascorbic acid in clinical analysis.