5-Bromo-5-nitro-1,3-dioxane, 98%, Thermo Scientific Chemicals
5-Bromo-5-nitro-1,3-dioxane, 98%, Thermo Scientific Chemicals
5-Bromo-5-nitro-1,3-dioxane, 98%, Thermo Scientific Chemicals
5-Bromo-5-nitro-1,3-dioxane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Bromo-5-nitro-1,3-dioxane, 98%, Thermo Scientific Chemicals
5-Bromo-5-nitro-1,3-dioxane, also known as bronidox, shows antimicrobial activity against gram-negative and gram-positive bacteria, yeast, and fungi.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFJ65443.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Water Content (Karl Fischer Titration)≤100 ppm
Appearance (Color)Colorless
Assay (GC)≥99.9%
Residue on Evaporation≤3 ppm
5-Bromo-5-nitro-1,3-dioxane is used as a stabilizer and preserving agent for biological molecules and solutions such as antibodies and antisera. Bronidox is used in a variety of rinse-off cosmetic. It can be used alone or in combination with methylisothiazolone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• 5-Bromo-5-nitro-1,3-dioxane is a chemical compound showing antimicrobial activity against gram-negative and gram-positive bacteria, yeast, and fungi
• This compound is capable of promoting the oxidation of essential protein thiols, causing inhibition of enzyme activity leading to the inhibition of microbial growth
Application
• 5-Bromo-5-nitro-1,3-dioxane has been used as a stabilizer and preserving agent for biological molecules and solutions, including antibodies and antisera
• It can be used alone or combined with methylisothiazolinone, which is also considered to be an effective preservative
RUO – Research Use Only
General References:
  1. Ya. Legin, G. 2-Bromo-2-nitro-1,3-propanediol(Bronopol) and its derivatives: Synthesis, properties, and application (a review). Pharmaceutical Chemistry Journal. 1996, 30, (4), 273-284.
  2. Ghannoum, M.; Thomson, M.; Bowman, W.; Al-Khalil, S. Mode of action of the antimicrobial compound 5-Bromo-5-nitro-1,3-dioxane (Bronidox). Folia Microbiologica. 1986, 31, (1), 19-31.