8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
Fluorescent probe for proteins on polyacrylamide gels
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALFJ65332.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS308080-99-1
IUPAC NameMolecular sieves 3A
MDL NumberMFCD00131613
SpecificationsSpecification SheetSpecification Sheet
Water adsorption>=21 % (50%rH, 25°C, 1bar)
Particle size>=95 % (3-5 mm)
Appearance (Color)Light grey to beige
Appearance (Form)Beads
Bulk density680 to 800 g/l
View more
Fluorescent probe for proteins on polyacrylamide gels