8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
8-Anilinonaphthalene-1-sulfonic acid, 95%, Thermo Scientific Chemicals
Fluorescent probe for proteins on polyacrylamide gels
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number ALFJ65332.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material8-Anilinonaphthalene-1-sulfonic acid
CAS82-76-8
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Fluorescent probe for proteins on polyacrylamide gels