Sucrose, Molecular Biology Grade
Sucrose, Molecular Biology Grade
Sucrose, Molecular Biology Grade
Sucrose, Molecular Biology Grade
Thermo Scientific Chemicals
Sucrose, Molecular Biology Grade
For protein purification by sucrose density gradients
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Catalog number ALFJ65148.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSucrose
CAS57-50-1
Melting Point185°C to 187°C
ColorWhite
Recommended StorageAmbient temperatures
View more
Sucrose is used to prepare density gradients for cell/organelle separation. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays. Sucrose is used in molecular biology for the preparation of density gradients.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sucrose is used to prepare density gradients for cell/organelle separation. In addition, sucrose can be used as a supplement in plant, insect, and bacterial culture media. It can also be used in various enzymatic assays. Sucrose is used in molecular biology for the preparation of density gradients.
Solubility
Soluble in water
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Ruby I. MacDonald. Temperature and ionic effects on the interaction of erythroid spectrin with phosphatidylserine membranes. Biochemistry. 1993, 32, (27),6957-6964.
  2. Henry Daniell. Transformation and Foreign Gene Expression in Plants Mediated by Microprojectile Bombardment. Recombinant Gene Expression Protocols. 1997, 62, 463-489.