(+/-)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
(+/-)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
(+/-)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
(+/-)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
Thermo Scientific Chemicals
(+/-)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
CAS: 74341-63-2 | C7H10N2O4 | 186.167 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ64884.MA10 mg
Catalog number ALFJ64884.MA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mg
Request bulk or custom format
Chemical Identifiers
CAS71924-62-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or solid
Infrared spectrumConforms
Melting point98°C to 100°C
GC>=96.0 %
Selective quisqualate agonist