Tropine DL-tropate, 99%, Thermo Scientific Chemicals
Tropine DL-tropate, 99%, Thermo Scientific Chemicals
Tropine DL-tropate, 99%, Thermo Scientific Chemicals
Tropine DL-tropate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tropine DL-tropate, 99%, Thermo Scientific Chemicals
Reduces brain levels of acetylcholine
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
Catalog number ALFJ64677.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS75-12-7
IUPAC Nameformamide
Molecular FormulaCH3NO
InChI KeyZHNUHDYFZUAESO-UHFFFAOYSA-N
SMILESNC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.05 % (Coulometric)
Sulfated ash=<0.005 %
Color scale=<10 APHA
GC>=99.5 %
Heavy metals (as Pb)=<1 ppm
View more
Tropine DL-tropate is used as a reduces brain levels of acetylcholine, Competitive nonselective antagonist at central and peripheral muscarinic acetylcholine receptors, used in opthalmology to dilate the pupil and in efforts of resuscitation in the treatment of bradycardia and cardiac arrest.