Salubrinal
Salubrinal
Salubrinal
Thermo Scientific Chemicals
Salubrinal
Have Questions?
Catalog NumberQuantity
ALFJ64192.MA10 mg
Catalog number ALFJ64192.MA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 mg
Request bulk or custom format
Specifications
Quantity10 mg
DescriptionPale Brown
Recommended StorageKeep cold
CAS405060-95-9
Molecular FormulaC21H17Cl3N4OS, C21H17Cl3N4OS
View more
Salubrinal is a selective Inhibitor of eIF2α (eukaryotic translation initiation factor 2 α-subunit) dephosphorylation, inhibiting global protein translation. Salubrinal is a specific inhibitor of ER Stress induced apoptosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
2°C to 8°C