Ammonium sulfate, 99+%, Molecular Biology Grade
Ammonium sulfate, 99+%, Molecular Biology Grade
Ammonium sulfate, 99+%, Molecular Biology Grade
Ammonium sulfate, 99+%, Molecular Biology Grade
Thermo Scientific Chemicals
Ammonium sulfate, 99+%, Molecular Biology Grade
CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ64180.A11 kg
Catalog number ALFJ64180.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS596-43-0
IUPAC Name(bromodiphenylmethyl)benzene
Molecular FormulaC19H15Br
InChI KeyNZHXEWZGTQSYJM-UHFFFAOYSA-N
SMILESBrC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)151-156?C
FormPowder or crystalline powder
Appearance (Color)White to yellow
Assay (Titration ex Bromide)≥97.5 to ≤102.5%