Cellulase, chromatographically purified, T. reesei
Cellulase, chromatographically purified, T. reesei
Thermo Scientific Chemicals
Cellulase, chromatographically purified, T. reesei
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number ALFJ64019.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Recommended StorageKeep cold
EINECS Number232-734-4
Quantity250 mg
FormLyophilized
Unit Size
Cellulase has been used to degrade cello-oligosaccharides into glucose and to study the biodegradability of bioabsorbable bacterial cellulose (BBC) material. It has been used in a study to assess the differential regulation of the cellulase transcription factors XYR1, ACE2, and ACE. It is also applied in the biodegradation of paper products such as foolscap paper, filter paper, newspaper and office paper as well as microcrystalline cellulose. It is added to detergents to improve cleansing properties.