Phenol, ultrapure, 99%, unstab.
Phenol, ultrapure, 99%, unstab.
Phenol, ultrapure, 99%, unstab.
Phenol, ultrapure, 99%, unstab.
Thermo Scientific Chemicals
Phenol, ultrapure, 99%, unstab.
CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ64011.36500 g
Catalog number ALFJ64011.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenol
Name Noteunstabilized
CAS108-95-2
Health Hazard 1H301+H311+H331-H314-H335-H341-H373
Health Hazard 2GHS H Statement
H301-H311-H331-H314-H341-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
View more
Phenol can be used for a wide variety of molecular biology applications, including nucleic acid extractions using standard phenol-chloroform protocols. Epidermal strips from the abaxial side of leaves of Commelina communis were stirred in phenol in a procedure to separate the monosaccharides from the epidermis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenol can be used for a wide variety of molecular biology applications, including nucleic acid extractions using standard phenol-chloroform protocols. Epidermal strips from the abaxial side of leaves of Commelina communis were stirred in phenol in a procedure to separate the monosaccharides from the epidermis.
Solubility
Soluble in water (82 g/L at 20°C).
Notes
Air sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only
General References:
  1. Sagit Shalel-Levanon; Ka-Yiu San; George N.Bennett. Effect of ArcA and FNR on the expression of genes related to the oxygen regulation and the glycolysis pathway in Escherichia coli under microaerobic growth conditions. Biotechnology and Bioengineering. 2005, 92,(2), 147-159.
  2. Louise Jones; Jennifer L.Milne; David Ashford; Simon J.McQueen-Mason. Cell wall arabinan is essential for guard cell function. PNAS. 2003, 100,(20), 11783-11788.