Phosphatase Inhibitor Cocktail I
Phosphatase Inhibitor Cocktail I
Thermo Scientific Chemicals
Phosphatase Inhibitor Cocktail I
Have Questions?
Catalog NumberQuantity
ALFJ63907.AA5 mL
Catalog number ALFJ63907.AA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Request bulk or custom format
Specifications
ColorColorless
Product TypePhosphatase Inhibitor Cocktail
Recommended StorageStore at -20°C
FormLiquid
Quantity5 mL
View more
Phosphatase Inhibitor Cocktail I is used to inhibitors with a broad specificity for serine, cysteine, and acid proteases, and aminopeptidases. It has been tested on a wide variety of cells and tissue extracts to isolate/maintain intact proteins without degradation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C