Agarose Gel Loading Dye (6X, Glycerol-Based)
Agarose Gel Loading Dye (6X, Glycerol-Based)
Thermo Scientific Chemicals
Agarose Gel Loading Dye (6X, Glycerol-Based)
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
50 mL
Catalog number ALFJ63869.AC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS19524-06-2
IUPAC Name4-bromopyridine hydrochloridyl
Molecular FormulaC5H4BrClN
InChI KeyPSPIMDWZDKGZBO-UHFFFAOYSA-N
SMILES[Cl].BrC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow-pink
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Form)Powder
Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.
Solubility
Fully miscible in water.
Notes
0.25% bromophenol blue, 0.25% xylene cyanol FF and 40% glycerol
RUO – Research Use Only
General References:
  1. Georges F. Carle and Maynard V. Olson. Separation of chromosomal DNA molecules from yeast by orthogonal-field-alternation gel electrophoresis. Nucleic Acids Research.1984, 12 (14), 5647-5664.
  2. Robert P. Noble. Electrophoretic separation of plasma lipoproteins in agarose gel. The Journal of Lipid Research.1968, 9 693-700.