Sodium phosphate, 0.5M buffer soln., pH 7.0
Sodium phosphate, 0.5M buffer soln., pH 7.0
Thermo Scientific Chemicals
Sodium phosphate, 0.5M buffer soln., pH 7.0
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
250 mL
Catalog number ALFJ63791.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium phosphate buffer soln.
Recommended StorageAmbient temperatures
Concentration0.5 M
TSCAYes
pH7.0
View more
Sodium phosphate is widely used in molecular biology, biochemistry, and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium phosphate is widely used in molecular biology, biochemistry, and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.
Solubility
Fully miscible.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
General References:
  1. Masters, C.L.; Simms, G.; Weinman, N.A.; Multhaup, G.; McDonald, B.L.; Beyreuther, K. Amyloid plaque core protein in Alzheimer disease and Down syndrome. Current Issue. 1985, 82, (12), 4245-4249.
  2. Volker M. Vogt. Purification and Further Properties of Single-Strand-Specific Nuclease from Aspergillus oryzae. European Journal of Biochemistry. 1973, 33, (1), 192-200.