Methyl caffeate, Thermo Scientific Chemicals
Methyl caffeate, Thermo Scientific Chemicals
Methyl caffeate, Thermo Scientific Chemicals
Methyl caffeate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl caffeate, Thermo Scientific Chemicals
Inhibitor of protein tyrosine kinase
Have Questions?
Catalog NumberQuantity
ALFJ63786.031 g
Catalog number ALFJ63786.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS5329-14-6
IUPAC Namesulfamic acid
Molecular FormulaH3NO3S
InChI KeyIIACRCGMVDHOTQ-UHFFFAOYSA-N
SMILESNS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point200°C to 208°C
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Inhibitor of protein tyrosine kinase