Potassium chloride, 1M aq. soln., Thermo Scientific Chemicals
Potassium chloride, 1M aq. soln., Thermo Scientific Chemicals
Potassium chloride, 1M aq. soln., Thermo Scientific Chemicals
Potassium chloride, 1M aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium chloride, 1M aq. soln., Thermo Scientific Chemicals
CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
Catalog number ALFJ63739.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS4420-74-0
IUPAC Name3-(trimethoxysilyl)propane-1-thiol
Molecular FormulaC6H16O3SSi
InChI KeyUUEWCQRISZBELL-UHFFFAOYSA-N
SMILESCO[Si](CCCS)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=80 %
Refractive index1.4410 to 1.4450 (20°C, 589 nm)
Potassium chloride 1M solution is used as conductive standard and as electrolyte in analytical chemistry.