TAE (10X), TRIS + acetate + EDTA
TAE (10X), TRIS + acetate + EDTA
Thermo Scientific Chemicals
TAE (10X), TRIS + acetate + EDTA
TAE (10X), TRIS + acetate + EDTA is a buffer composed of Tris, acetate, and EDTA components and is used in electrophoresis of nucleic acids in agarose and polyacrylamide gels.
Have Questions?
Catalog NumberQuantity
ALFJ63677.K21 L
Catalog number ALFJ63677.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialTAE (10X)
Recommended StorageAmbient temperatures
TSCAYes
Physical FormLiquid
Quantity1 L
View more
TAE is used for DNA agarose gel electrophoresis but is also used for non-denaturing RNA agarose gel electrophoresis, used at various concentrations to study the mobility of DNA in solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • TAE (10X), TRIS + acetate + EDTA allows nucleic acid separation
Application
  • This buffer has been used to prepare agarose and polyacrylamide gels for electrophoresis of nucleic acids
  • It is used as a running and gel preparation buffer, and it is also used for non-denaturing RNA agarose gel electrophoresis
RUO – Research Use Only
General References:
  1. Lechanteur, A.; Furst, T.; Evrard, B.; Delvenne, P.; Hubert, P.; Piel, G. Development of anti-E6 pegylated lipoplexes for mucosal application in the context of cervical preneoplastic lesions. Int. J. Pharm. 2015, 483 (1-2), 268-277.
  2. Kaur, K.; Kumar, V.; Beniwal, V.; Kumar, V.; Aneja, K. R.; Sharma, V.; Jaglan, S. T. Solvent-free synthesis of novel (E)-2-(3, 5-dimethyl-4-(aryldiazenyl)-1H-pyrazol-1-yl)-4-arylthiazoles: determination of their biological activity. Med. Chem. Res. 2015, 24 (11), 3863-3875.