Semi dry blot transfer buffer (10X)
Semi dry blot transfer buffer (10X)
Thermo Scientific Chemicals
Semi dry blot transfer buffer (10X)
Have Questions?
Catalog NumberQuantity
ALFJ63664.K21L
Catalog number ALFJ63664.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1L
Request bulk or custom format
Chemical Identifiers
CAS126-33-0
IUPAC Name1λ⁶-thiolane-1,1-dione
Molecular FormulaC4H8O2S
InChI KeyHXJUTPCZVOIRIF-UHFFFAOYSA-N
SMILESO=S1(=O)CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4805-1.4850 @ melt
FormLiquid as melt
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤0.5%
Appearance (Color)Clear colorless to yellow