Etoposide, Thermo Scientific Chemicals
Etoposide, Thermo Scientific Chemicals
Etoposide, Thermo Scientific Chemicals
Etoposide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Etoposide, Thermo Scientific Chemicals
Topoisomerase inhibitor and inducer of apoptosis
Have Questions?
Catalog NumberQuantity
ALFJ63651.MB25 mg
Catalog number ALFJ63651.MB
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mg
Request bulk or custom format
Chemical Identifiers
CAS106-38-7
IUPAC Name1-bromo-4-methylbenzene
Molecular FormulaC7H7Br
InChI KeyZBTMRBYMKUEVEU-UHFFFAOYSA-N
SMILESCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting crystalline mass
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)White
Etoposide is used as a inhibits DNA synthesis and is very active against cells in the late S and G2 phases of the cell cycle. Etoposide was reported to be used to serum starve Saos-2 M5(p53-/-) cells which were transfected by electroporation with indicated expression plasmids for flag-E2F1 and myc-EBNA3C for selection of colony forming units, thus studying the blocking effect of EBV Nuclear Antigen 3C on E2F1 Mediated Apoptosis Induced by the DNA Damage Response.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Etoposide is used as a inhibits DNA synthesis and is very active against cells in the late S and G2 phases of the cell cycle. Etoposide was reported to be used to serum starve Saos-2 M5(p53-/-) cells which were transfected by electroporation with indicated expression plasmids for flag-E2F1 and myc-EBNA3C for selection of colony forming units, thus studying the blocking effect of EBV Nuclear Antigen 3C on E2F1 Mediated Apoptosis Induced by the DNA Damage Response.
Solubility
Insoluble in water.
Notes
Store solutions in DMSO at 4°C. For long term storage prepare aliquots and store at -20°C.
RUO – Research Use Only
General References:
  1. Hande, K.R. Etoposide: four decades of development of a topoisomerase II inhibitor.European Journal of Cancer.1998, (10), 1514-1521.
  2. Maurice L. Slevin. The clinical pharmacology of etoposide.Supplement: Recent Advances in Etoposide Therapy Combined Proceedings of Two Symposia.1991, (1), 319-329.