Thiamphenicol, Thermo Scientific Chemicals
Thiamphenicol, Thermo Scientific Chemicals
Thiamphenicol, Thermo Scientific Chemicals
Thiamphenicol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiamphenicol, Thermo Scientific Chemicals
The methyl-sulfonyl analogue of chloramphenicol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFJ63575.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1073-13-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥95.0% (non-U.S. specification)
Assay from Suppliers CofA≥95.0% (GC, U.S. specification)
CommentPurchased in the U.S. and in other countries
View more
Thiamphenicol is used to treat chancroid in men and uncomplicated gonorrhea. It is used in studies of bacterial protein synthesis at the level of peptidyl transferase activity associated with the 23S rRNA of the 50S ribosomal subunit. It is used to study chloraniphenicol-thiamphenicol-resistance and the use of fluorinated analogs when resistance is encountered. Its main advantage over chloramphenicol is that it has never been associated with aplastic anaemia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiamphenicol is used to treat chancroid in men and uncomplicated gonorrhea. It is used in studies of bacterial protein synthesis at the level of peptidyl transferase activity associated with the 23S rRNA of the 50S ribosomal subunit. It is used to study chloraniphenicol-thiamphenicol-resistance and the use of fluorinated analogs when resistance is encountered. Its main advantage over chloramphenicol is that it has never been associated with aplastic anaemia.
Solubility
Soluble in acetonitrile or DMF. Slightly soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only
General References:
  1. Tomoko Nagata.; Hisao Oka. Detection of Residual Chloramphenicol, Florfenicol, and Thiamphenicol in Yellowtail Fish Muscles by Capillary Gas Chromatography-Mass Spectrometry.J. Agric. Food Chem.1996, (5), 1280-1284.
  2. Franklin A Davis.; Ping Zhou. Asymmetric synthesis of the antibiotic (+)-thiamphenicol using cis-N-(p-toluenesulfinyl)aziridine 2-carboxylic acids.Tetrahedron Letters.1994, (41), 7525-7528.