Cetirizine dihydrochloride, 99+%, Thermo Scientific Chemicals
Cetirizine dihydrochloride, 99+%, Thermo Scientific Chemicals
Cetirizine dihydrochloride, 99+%, Thermo Scientific Chemicals
Cetirizine dihydrochloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cetirizine dihydrochloride, 99+%, Thermo Scientific Chemicals
CAS: 83881-52-1 | C21H27Cl3N2O3 | 461.81 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ63549.065 g
Catalog number ALFJ63549.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCetirizine dihydrochloride
CAS83881-52-1
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
Cetirizine dihydrochloride is used as a Histamine H-1 receptor antagonist, antihistaminic, piperazines. Inhibits activation of eosinophils and neutrophils. Non-sedating type histamine H1-receptor antagonist; major metabolite of hydroxyzine. Pharmacological activity resides primarily in the (R)-isomer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cetirizine dihydrochloride is used as a Histamine H-1 receptor antagonist, antihistaminic, piperazines. Inhibits activation of eosinophils and neutrophils. Non-sedating type histamine H1-receptor antagonist; major metabolite of hydroxyzine. Pharmacological activity resides primarily in the (R)-isomer.
Solubility
Soluble in water, DMSO, ethanol, and methanol.
Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only