Stacking buffer (4X)
Stacking buffer (4X)
Thermo Scientific Chemicals
Stacking buffer (4X)
Have Questions?
Catalog NumberQuantity
ALFJ63450.K21 L
Catalog number ALFJ63450.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS9003-39-8
Molecular Formula(C6H9NO)n
SMILES*-CC(-*)N1CCCC1=O
MDL NumberMFCD01076626
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Elemental AnalysisNitrogen: 11.5-12.8%
Water Content (Karl Fischer Titration)≤5%
pH3.0-7.0
Assay (special)K value: 27-32
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Ambient Temperatures